missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Resurge™ CD4 Supplement, without glucose and L-glutamine
Description
A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. |
| Cell Line | CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free |
| Culture Type | Mammalian Cell Culture |
| Form | Powder |
| Product Type | Media Supplement |
| Without Additives | No Glutamine |
| Format | Bottle(s) |
| Product Line | Resurge |
| Quantity | 100 g |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?