missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD5 Supplement, without glucose and L-glutamine

Product Code. 16229921
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16229921 5 kg 5kg
16219921 1 kg 1kg
16209921 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16229921 Supplier Gibco™ Supplier No. 670026

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.

A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Culture Type Mammalian Cell Culture
Form Powder
Product Type Media Supplement
Without Additives No Glutamine
Format Bottle(s)
Product Line Resurge
Quantity 5 kg
Source Animal origin-free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.