missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Resuspension Solution for GeneJET Plasmid Miniprep Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ GeneJET Resuspension Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat No. K0502 and K0503) and may be purchased separately.
Highlights
- The Resuspension Solution should be stored at 4°C and is stable for 6 months.
- After the addition of RNase A, the Resuspension Solution is stable for 6 months when stored at 4°C.
Related Products
- Neutralization Solution for GeneJET Plasmid Miniprep Kit (Cat. No. R1233)
- Wash Solution for GeneJET Plasmid Miniprep Kit (R1243)
- Binding Buffer for GeneJET Plasmid Miniprep Kit (R1331)
- GeneJET NGS Cleanup Kit (K0851)
- Binding Buffer for GeneJET NGS Cleanup Kit (R2811)
- Prewash Buffer for GeneJET NGS Cleanup Kit (R2821)
- Wash Buffer for GeneJET NGS Cleanup Kit (R2831)
- GeneJET Plasmid Miniprep Kit (K0502)
- RNase A (R1253)
- Elution Buffer for GeneJET Plasmid Miniprep Kit (R1263)
- Lysis Solution for GeneJET Plasmid Miniprep Kit (R1223)
Specifications
Specifications
| Buffer | Suspension Buffer |
| Content And Storage | Store at room temperature,-20°C |
| Description | Reagent |
| Format | Kit |
| Product Line | GeneJET |
| Product Type | Resuspension Solution |
| Quantity | 70 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?