missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Resveratrol

Product Code. 17100142
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 17100142

Brand: STEMCELL Technologies 72864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antioxidant; COX-1 inhibitor

  • Resveratrol is a potent phenolic antioxidant found in grapes and red wine that also has antiproliferative and anti-inflammatory activity (Rotondo et al.) Resveratrol is also a selective inhibitor of cyclooxygenase-1 (COX-1; Jang) with an ED50 value of 15 μM, respectively
  • Inhibits differentiation of pig preadipocytes into adipocytes (Bai et al.)
  • Promotes self-renewal of human mesenchymal stem cells (Peltz et al.)
  • Induces osteogenic differentiation in human bone marrow-derived mesenchymal stem cell cultures (Boissy et al.; Dai et al.; Peltz et al.)
  • Induces osteoblast differentiation while blocking adipocyte development in mouse mesenchymal stem cells and cultured primary rat bone marrow cells (Bäckesjö et al.)

Specifications

CAS 501-36-0
Synonym (E)-Resveratrol; trans-Resveratrol
Quantity 500 mg
Molecular Formula C14H12O3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.