missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Resveratrol
GREENER_CHOICE

Product Code. 30001250
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30001250

Brand: Tocris Bioscience™ 1418/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cyclooxygenase inhibitor

A phytoestrogen with antitumor, antioxidant, antiplatelet, anti-inflammatory and antifungal effects. Inhibits cytochrome P450 1A1 (IC50 = 23 μM) and displays mixed agonist/antagonist actions at ERα and ERβ estrogen receptors. Converted into the anticancer agent piceatannol (Cat. No. 1554) by cytochrome P450 1B1. Activates autophagy. Also activates TRPA1 in prostate cancer-associated fibroblasts. Also SIRT1 activator.
TRUSTED_SUSTAINABILITY

Specifications

CAS 501-36-0
Quantity 100 mg
Molecular Formula C14H12O3
Formula Weight 228.25
Purity >98%
Molecular Weight (g/mol) 228.247
Chemical Name or Material Resveratrol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.