missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Allows parameters and process data to be viewed and adjusted without working directly on the device, reducing contact with hazardous media and minimizing the risk of accidents
- Provides greater safety through operation outside the fume hood
- The included docking station enables permanent mounting, for example, directly on the outside of the fume hood
- Ensures the fume hood remains closed throughout the entire experiment while providing access to all relevant information at all times
- This offers crucial safety benefits for:
- Working with toxic, volatile or flammable substances
- High-temperature applications
- Long-term or unattended operation
- Provides central control of multiple devices
- Enables up to five devices to be read out and controlled simultaneously
- Simplifies monitoring in more complex experimental setups and supports structured, reproducible laboratory workflows
Specifications
Specifications
| Top Plate Material | Aluminum Alloy |
| Voltage | 220/230 V |
| Size | 135 mm dia. |
| Temperature (Metric) | 0°C to 340°C |
| Type | Magnetic Stirrer With Remote Control And Monitoring |
| Plug Type | UK |
| No. of Positions | 1 |
| Max. Container Size | 20 L |
| Stirring Range | 50 to 1700 rpm |
| Warranty | 2 Years |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?