missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500 Dx Series 8-Tube Standard Retainer & Base Set

Product Code. 16281041 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Sets
Unit Size:
4 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16281041 4 Sets 4 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16281041 Supplier Applied Biosystems™ Supplier No. 4410230
  • ONLINE EXCLUSIVE PRICE
    364.00 €
    309.40 € / 4 sets
    Save 54.60 €
    15% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is an 8-tube retainer & base set (standard) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.

This is a retainer and base set (standard) designed for septa retention in 8-tube strip format for use with the 3500 Dx and 3500xL Dx genetic analyzers.

Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only. Not for use in the USA.

  • This is an 8-tube retainer & base set (standard) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Ambient storage.
Each package contains 4 sets.
Format 8-tube Strip
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Retainer and Base Set
Quantity 4 Sets
Shipping Condition Room Temperature

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.