missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Retainer & Base Set (Standard) for 3500/3500xL Genetic Analyzers, 8 tube

Product Code. 15879846 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Sets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879846 4 Sets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15879846 Supplier Applied Biosystems™ Supplier No. 4410231
  • ONLINE EXCLUSIVE PRICE
    373.00 €
    317.05 € / Pack of 1
    Save 55.95 €
    15% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This 8-tube retainer and base set is for use with standard 8-tube strips on the 3500 series Genetic Analyzers.

This 8-tube retainer and base set is for use with standard 8-tube strips on the 3500 series Genetic Analyzers.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This product includes 4 sets of retainers and bases.
Description Retainer & Base Set (Standard), 8-well Strips, 0.2mL, For 3500/3500XL Genetic Analyzers, Room Temperature, Standard Reaction Speed
Format 8-well Strip
For Use With (Equipment) 3500 Genetic Analyzer, 3500xL Genetic Analyzer
For Use With (Application) Sequencing
Product Type Base Set
Quantity 4 Sets
Sample Volume 0.2 mL
Shipping Condition Room Temperature
Test Time Standard
Tube Type Strip Tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.