missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Retainer Ring

Product Code. p-7162774
Click to view available options
Color:
Chrome
Gray
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10044270

Brand: Hirschmann™ 9903802

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Spare part retainer ring chromated for pipetus™ pipetholder.

  • Spare part retainer ring chromated for pipetus™ pipetholder.

Specifications

Color Chrome
Type Ring Union Nut
For Use With (Equipment) Pipetus, Pipetus Junior, and Pipetus Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.