missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Retinoic acid

Product Code. 30243662
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30243662 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30243662 Supplier R&D Systems™ Supplier No. TB0695RMU/50
  • ONLINE EXCLUSIVE PRICE
    500.30 €
    452.55 € / 50mg
    Save 47.75 €
    9% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Retinoic acid synthesized to Ancillary Material Grade

TB0695-RMU is an Ancillary Material Grade version of Retinoic acid (Cat. No. 0695). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2E,4E,6E,8E-nonatetraenoic acid
CAS 302-79-4
Quantity 50 mg
Product Line Tocris
Target Retinoic Acid Receptor Agonists
Molecular Formula C20H28O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.