missing translation for 'onlineSavingsMsg'
Learn More

Retinoic Acid, All Trans Isomer, MP Biomedicals™

Product Code. 11431771 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431771 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431771 Supplier MP Biomedicals Supplier No. 02190269.6
  • ONLINE EXCLUSIVE PRICE
    55.35 €
    52.50 € / 50mg
    Save 2.85 €
    5% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mediates the functions of vitamin A required for growth and development. Retinoic Acid, All Trans Isomer, MP Biomedicals has been used in various differentiation protocols, including B-cells, T cells and neurons, and is applied clinically to treat cancer

Retinoic acid (RA) is a vitamin A-derived, non-peptidic, small lipophilic molecule. It affects gene transcription and modulates a wide variety of biological processes, including cell proliferation and differentiation, including apoptosis.

  • Retinoic Acid mediated gene transcription depends on the rate of transport of Retinoic Acid to target cells and the timing of exposure of Retinoic Acid to RARs (Retinoic Acid Receptors) in the target tissues.
  • All-trans-Retinoic acid (ATRA) is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR).
  • The bound RAR and RXR act as transcription factors that regulate the growth and differentiation of both normal and malignant cells.
  • Cytochromes P450 (CYPs) catalyze the 4-hydroxylation of ATRA.
  • Retinoic acid primes embryonic stem cells to become neurons.

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
MDL Number MFCD00001551
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
Synonym retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra
PubChem CID 444795
ChEBI CHEBI:15367
SMILES C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O

Specifications

Melting Point 173°C to 183°C
Quantity 50 mg
Solubility Information Soluble in methanol, chloroform (40mg/mL) or acetone (30mg/mL), slightly soluble in ethanol.
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
Formula Weight 300.4
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • Retinoic Acid, All Trans Isomer
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.