missing translation for 'onlineSavingsMsg'
Learn More

Retinol, 95%, synthetic

Product Code. 15434529
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434529 1 g 1g
15444529 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434529 Supplier Thermo Scientific Alfa Aesar Supplier No. J62079.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 68-26-8 | C20H30O | 286.46 g/mol

It induces cell differentiation and apoptosis of some cancer cell types and inhibiting carcinogenesis. It plays an essential role in metabolic functioning of the retina, the growth of and differentiation of epithelial tissue, the growth of bone, reproduction, and the immune response. It acts as a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) that act as transcription factors to regulate the growth and differentiation of normal and malignant cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-26-8
Molecular Formula C20H30O
MDL Number MFCD00001552
InChI Key FPIPGXGPPPQFEQ-OVSJKPMPSA-N
PubChem CID 445354
ChEBI CHEBI:17336

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 403040
Sensitivity Light and moisture sensitive
Merck Index 14,10013
Synonym retinol, vitamin a, all-trans-retinol, vitamin a1, alphalin, axerophthol, vitamin a alcohol, oleovitamin a, chocola a, alphasterol
Solubility Information Practically insoluble in water or glycerol; soluble in absolute alcohol, methanol, chloroform,ether,fats and oils.
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
Molecular Weight (g/mol) 286.459
Formula Weight 286.46
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material Retinol, synthetic
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Retinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Vitamin A

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.