missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Retinol Binding Protein ELISA Kit

Product Code. p-200006767
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17104863

Brand: Invitrogen™ EIARBP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Retinol Binding Protein Competitive ELISA quantitates retinol binding protein (RBP) in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The Retinol Binding Protein Competitive ELISA research-use-only kit is designed to quantitatively measure RBP independent of species. An RBP standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-sheep IgG antibodies. Anti-sheep RBP antibodies and RBP-peroxidase conjugate is added to the wells. The RBP-peroxidase conjugate and any RBP in the sample will compete to bind to the anti-sheep RBP antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound RBP-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of RBP present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; Rbp1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellular
Assay Range 7.8 to 1000 ng/mL
Assay Sensitivity 1.36 ng/mL, 1.36 μg/mL, 1.36 μg/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5947
Gene Symbol Rbp1
Interassay CV 12%
Intraassay CV 5
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target RBP1
Test Time 45 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.