missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Retinyl acetate

Product Code. 16439126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439126 Supplier MedChemExpress Supplier No. HYN0679
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Retinyl acetate is a synthetic acetate ester form derived from retinol and has potential antineoplastic and chemo preventive activities.

Specifications

Content And Storage Pure form -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 328.49 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 127-47-9
Product Type Natural Products
Purity 99.29%
Solubility DMSO : 50 mg/mL (152.21 mM∣Need ultrasonic) ∣H2O : 0.67 mg/mL (2.04 mM∣Need ultrasonic)
Synonym Retinol acetate∣ Vitamin A acetate
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.