Learn More
Erlab Retractable Shelf for Adjustable Base
Product Code. 31794097 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Retractable Shelf
Unit Size:
Each
Product Code. Product Type Unit Size
31794097 Retractable Shelf Each
1 options
Product Code. 31794097 Supplier Erlab Supplier No. NORT000399
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
Designed for adjustable legs, this retractable shelf allows you to add a functional storage surface whilst retaining a high degree of flexibility in its use.
  • Benches, Rolling Carts and Bases for Erlab Enclosures: Erlab benches and rolling carts are designed to supPort ductless enclosures and filtering cabinets at the correct working height, in complete safety. Each model is dimensioned for a specific enclosure reference to guarantee stability and a perfect match with the equipment footprint.
  • Stability and Ergonomics:
    • Robust steel structure sized for the load of the corresponding enclosure
    • Chemically resistant, easy-to-clean finish
    • Fixed, mobile, levelling-feet, mechanically adjustable and electrically adjustable versions available
    • Height-adjustable models allow the working position to be adapted to each operator, seated or standing
    • Lockable castors on rolling versions for safe repositioning of the workstation
  • Flexible Laboratory Layout: Rolling carts make it possible to move a complete filtration workstation from one room to another as projects evolve, while fixed benches provide a permanent, space-efficient installation. Optional accessories such as retractable shelves and footrests further improve daily comfort and storage capacity.
  • Key Benefits:
    • Dedicated supPort designed for a specific Erlab enclosure.
    • Stable, robust structure with chemically resistant finish.
    • Fixed, mobile and height-adjustable versions available.
    • Improved operator ergonomics and workstation flexibility.
    • Simple assembly and easy maintenance.
Specifications
Product Type Retractable Shelf