missing translation for 'onlineSavingsMsg'
Learn More

Revefenacin, MedChemExpress

Product Code. 16440266
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440266

Brand: Medchem Express HY15851

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Revefenacin (TD-4208; GSK1160724) is a potent mAChR antagonist; has a high affinity on M3 receptor with a Ki of 0.18 nM.

Revefenacin (TD-4208

Specifications

Chemical Name or Material Revefenacin
CAS 864750-70-9
Color White
Purity Grade Notes Research
Molecular Formula C35H43N5O4
Quantity 2 mg
Synonym TD-4208 GSK1160724
Solubility Information DMSO : 100 mg/mL (167.29 mM; Need ultrasonic)
SMILES O=C(OC1CCN(CCN(C(C2=CC=C(CN3CCC(C(N)=O)CC3)C=C2)=O)C)CC1)NC4=CC=CC=C4C5=CC=CC=C5
Molecular Weight (g/mol) 597.75
Formula Weight 597.75
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.