missing translation for 'onlineSavingsMsg'
Learn More

Revefenacin, MedChemExpress

Product Code. 16489407
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489407 5 mg 5mg
16452937 50 mg 50mg
16446966 100 mg 100mg
16440266 2 mg 2mg
16406878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489407 Supplier Medchem Express Supplier No. HY15851
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Revefenacin (TD-4208; GSK1160724) is a potent mAChR antagonist; has a high affinity on M3 receptor with a Ki of 0.18 nM.

Revefenacin (TD-4208

Specifications

Chemical Name or Material Revefenacin
CAS 864750-70-9
Color White
Purity Grade Notes Research
Molecular Formula C35H43N5O4
Quantity 5 mg
Synonym TD-4208 GSK1160724
Solubility Information DMSO : 100 mg/mL (167.29 mM; Need ultrasonic)
SMILES O=C(OC1CCN(CCN(C(C2=CC=C(CN3CCC(C(N)=O)CC3)C=C2)=O)C)CC1)NC4=CC=CC=C4C5=CC=CC=C5
Molecular Weight (g/mol) 597.75
Formula Weight 597.75
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.