missing translation for 'onlineSavingsMsg'
Learn More

Revefenacin, TRC

Product Code. 30426536
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426536 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426536 Supplier Toronto Research Chemicals Supplier No. TRCR279600250MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Revefenacin
CAS 864750-70-9
Molecular Formula C35 H43 N5 O4
Quantity 250 mg
Synonym Carbamic acid','N-[1,1'-biphenyl]-2-yl-','1-[2-[[4-[[4-(aminocarbonyl)-1-piperidinyl]methyl]benzoyl]methylamino]ethyl]-4-piperidinyl ester, Carbamic acid','[1,1'-biphenyl]-2-yl-','1-[2-[[4-[[4-(aminocarbonyl)-1-piperidinyl]methyl]benzoyl]methylamino]ethyl]-4-
InChI Formula InChI=1S/C35H43N5O4/c1-38(34(42)29-13-11-26(12-14-29)25-40-19-15-28(16-20-40)33(36)41)23-24-39-21-17-30(18-22-39)44-35(43)37-32-10-6-5-9-31(32)27-7-3-2-4-8-27/h2-14,28,30H,15-25H2,1H3,(H2,36,41)(H,37,43)
SMILES CN(CCN1CCC(CC1)OC(=O)Nc2ccccc2c3ccccc3)C(=O)c4ccc(CN5CCC(CC5)C(=O)N)cc4
IUPAC Name [1-[2-[[4-[(4-carbamoylpiperidin-1-yl)methyl]benzoyl]-methylamino]ethyl]piperidin-4-yl] N-(2-phenylphenyl)carbamate
Molecular Weight (g/mol) 597.747
Formula Weight 597.332
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.