Learn More
Texwipe™ Revolve™ Mini AlphaMop™ Covers
Product Code. 17426613
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
17426613 Sterile Pack of 100
17476613 Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17426613 Supplier Texwipe™ Supplier No. STX1720
  • ONLINE EXCLUSIVE PRICE
    1390.00 €
    1144.70 € / Pack of 100
    Save 245.30 €
    17% Off
Estimated Shipment: 16-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Revolve™ Mini AlphaMop™ 2-in-1 Covers, the first sustainable covers for the cleanroom industry, are made from 100% upcycled polyester. Revolve™ mop covers aids in sustainability goals, reducing the amount of post-consumer water bottles going into landfills and carbon emissions.
  • Mopping and cleaning large and small surfaces.
  • Applying and removing disinfectants.
  • Spill control.
  • Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers.
  • Appropriate for use with temperatures less than 400°F (205°C).
  • The integrated polyester cover/pad design allows replacing covers and pads in a one-step process.
  • The integrated design offers high sorption capacity, which is ideal for spill control, cleaning, and solution application.
  • The covers easily fit over the mop head, permitting a quick change.
  • The integrated polyester pad assures the mop cover conforms to surfaces for consistent contact and cleaning.
  • Revolve™ processing provides low levels of ions, NVRs (non-volatile residues), particles and fibers for use in critical cleaning applications and environments.
  • Autoclave safe.
  • Sterile covers are individually lot coded for traceability and quality control.
Specifications
Product Type Mop Cover
Color Code White
Sterility Sterile
Autoclavable Autoclavable
Color-coded No
Material Polyester