Learn More
Invitrogen™ RFP Tag (aa 22-36) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP260
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
RFP tag (residues 22-36) Synthetic Peptide, PEP-260, from Invitrogen.
PEP-260 is a 15 amino acid synthetic peptide whose sequence is derived from the RFP tag protein. The sequence of this peptide is (amino to carboxy terminus): V (22)-N-G-H-E-F-E-I-E-G-E-G-E-G-R (36)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, RFP Epitope Tag, catalog # PA1-986. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
Non-species Specific | |
RFP Tag (aa 22-36) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
No preservative | |
50 μg | |
V(22) - N - G - H - E - F - E - I - E - G - E - G - E - G - R (36) |
purified | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Yes | |
Peptide | |
RUO - research use only |