missing translation for 'onlineSavingsMsg'
Learn More

RG3039, MedChemExpress

Product Code. 16462826
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462826 25 mg 25mg
16454766 100 mg 100mg
16430077 50 mg 50mg
16423688 10 mg 10mg
16416427 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16462826 Supplier Medchem Express Supplier No. HY102020
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.

RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.

Specifications

Chemical Name or Material RG3039
CAS 1005504-62-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23Cl2N5O
Quantity 25 mg
Synonym PF-06687859
Solubility Information DMSO : 5 mg/mL (11.56 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC1=NC(N)=C2C(OCC3CCN(CC4=C(Cl)C=CC=C4Cl)CC3)=CC=CC2=N1
Molecular Weight (g/mol) 432.35
Formula Weight 432.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.