missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHD Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | RHD |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RHD Polyclonal antibody specifically detects RHD in Mouse, Rat samples. It is validated for Western BlotSpecifications
| RHD | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, CD Markers, Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 6007 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| blood group Rh(D) polypeptide, CD240D, DIIIc, RH, Rh blood group, D antigen, RH30, Rh4, RHCED, RhDCw, RHDel, RHDVA(TT), RhII, RhK562-II, RhPI, RHPII, RHXIII | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human RHD (NP_001121163.1). LGNLRMVISNIFNTDYHMNMMHIYVFAAYFGLSVAWCLPKPLPEGTEDKDQTATIPSLSAMLGALFLWMFWPSFNSALLRSPIERKNAVFNTYYAVAVSVV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title