missing translation for 'onlineSavingsMsg'
Learn More

Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis)

Product Code. 11450390
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450390 50 x 50 mm Each
11331499 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11450390 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 013983.FI
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-15-5 | Re | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈6.58g/25x25mm
Form Foil
Percent Purity 99.97%
Assay (metals basis)
Chemical Name or Material Rhenium foil, 0.5mm (0.02 in.) thick, Annealed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.