Learn More
Thermo Scientific™ Rheodyne™ 8125 Low-Dispersion Microscale Injector
Product Code. 10505235 Shop All Thermo Scientific Products
Click to view available options
Mode:
Dual Mode
Unit Size:
Each
Product Code. 10505235
Brand: Thermo Scientific™ 8125
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure highly accurate and precise injections for microscale LC analysis with the Thermo Scientific™ Rheodyne™ 8125 Low-Dispersion Microscale Injector.
Ensure highly accurate and precise injections for 1 and 2mm I.D. high-performance liquid chromatography (HPLC) columns using the Thermo Scientific™ Rheodyne™ 8125 Low-Dispersion Microscale Injector. This dual-mode injector can use partial filling for zero sample waste or complete filling for better reproducibility. A built-in position-sensing switch provides a reproducible start signal. It is suitable for use with 5 to 50μL sample loops.
  • Can use partial filling for zero sample waste or complete filling for better reproducibility
  • Position sensing switch provides reproducible start signal
  • Suitable for use with 5 to 50μL sample loops
Includes:
5μL sample loop
TRUSTED_SUSTAINABILITY
Specifications
Mode Dual Mode
Volume (Metric) Sample 0.1 to 5 μL
Description Dual Mode; Stainless Steel Stator; Low Dispersion Micro-Scale; Position Sensing Switch
For Use With (Equipment) HPLC
Type Low Dispersion Injector
No. of Ports 6 Ports
Stator Material Stainless Steel
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.