missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ rhLaminin-521

Product Code. 15481044 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 mg
100μg
Unit Size:
100µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481044 1 mg 100µg
15461945 100μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481044 Supplier Gibco™ Supplier No. A29249
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

rhLaminin-521

Recombinant human Laminin 521 (rhLaminin-521) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8™ Medium, rhLaminin-521 has been proven to promote cellular survival under stressful conditions in the absence of small molecule inhibitors. rhLaminin-521 has been shown to support PSC growth for >20 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages. rhLaminin-521 also provides optimum reprogramming efficiency of human somatic cells as well as human pluripotent stem cell survival during feeder-dependent to feeder-free transitions when used in conjunction with Essential 8 Medium. For a kit that combines the medium and substrate, see the Essential 8™ Adaptation Kit.

rhLaminin-521 is:
  • Robust—transition cells between feeder-dependent and feeder-free culture systems with minimal effort
  • Efficient—easily reprogram human fibroblasts to pluripotent stem cells
  • Physiologically relevant—expand cells with minimal differentiation
  • Scalable—passage stem cell lines as single cells with no impact to genetic stability

    Trouble-free transition
    When used with Essential 8 Medium, rhLaminin-521 provides a robust culture matrix that supports pluripotent stem cell adherence and expansion during the transition from feeder-dependent culture systems.

    Efficient reprogramming
    rhLaminin-521 has been shown to increase the number of colonies obtained during reprogramming of human fibroblasts, which can be important for in vitro disease modeling.

    Confident expansion
    rhLaminin-521 is expressed in blastocysts and therefore provides an environment mimicking the human stem cell niche.

    Easy scale-up
    rhLaminin-521 promotes stable pluripotent stem cell maintenance as single cells in the absence of ROCK inhibitors.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-10 to -30°C)
    Cell Type Stem Cells
    Classification Xeno-free
    Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
    Product Type Extracellular Matrix
    pH 6 to 8
    Purity or Quality Grade hESC-Qualified
    Quantity 1 mg
    Shipping Condition Dry Ice
    Source Xeno-Free

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.