missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 110, 98+%, pure, laser grade

Product Code. 11966931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11966931 2.5 g Glass bottle 2.5g
10730682 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966931 Supplier Thermo Scientific Chemicals Supplier No. 419070025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13558-31-1 | C20H15ClN2O3 | 366.801 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13558-31-1
Molecular Formula C20H15ClN2O3
Molecular Weight (g/mol) 366.801
InChI Key JNGRENQDBKMCCR-UHFFFAOYSA-N
PubChem CID 11617572
IUPAC Name 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride
SMILES C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl

Specifications

Melting Point >360.0°C
Fluorescence Max 520nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 498 to 504nm (c=10mg/l in methanol)
Molar Absorption Coefficient 77000 l/(mol.cm) (at lambda max.) min.
Packaging Glass bottle
Suitability (for use as a laser dye) passes
Assay Percent Range 98+%
Color White to Yellow
Solubility Information Solubility in water: slightly soluble.
Quantity 2.5 g
Physical Form Crystalline Powder
Formula Weight 366.79
Percent Purity ≥98%
Grade Pure
Chemical Name or Material Rhodamine 110
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.