missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine 110, TRC

Product Code. 30427571
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427571 1 g
30424824 100 mg
30413235 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427571 Supplier Toronto Research Chemicals Supplier No. TRCR3185801G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13558-31-1
Molecular Formula C20 H14 N2 O3 . Cl H
Molecular Weight (g/mol) 366.8
Synonym 3,6-Diamino-9-(2-carboxyphenyl)xanthylium Chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic Acid Monohydrochloride, 3,6-Diamino-9-(2-carboxyphenyl)xanthylium Chloride, R 110, RH 110, Rhodamine 560, Rhodamine N
IUPAC Name 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride
SMILES Cl.Nc1ccc2C(=C3C=CC(=N)C=C3Oc2c1)c4ccccc4C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H14N2O3.ClH/c21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24;/h1-10,21H,22H2,(H,23,24);1H
Formula Weight 366.0771
Chemical Name or Material Rhodamine 110
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.