missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Rhodamine 123

Product Code. 16498758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498758 10 mg 10mg
16483816 25 mg 25mg
16474677 50 mg 50mg
16448906 100 mg 100mg
16441178 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498758 Supplier MedChemExpress Supplier No. HYD0816
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rhodamine 123 (RH-123; R-22420) is a fluorescent dye (λex=503 nm, λem=527 nm).

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 100 mg/mL (262.59 mM; Need ultrasonic)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 380.82 g/mol
CAS 62669-70-9
Synonym RH-123 ∣ R-22420
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.