missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 123, 99+%, pure

Product Code. p-190909 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 10578132

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62669-70-9 | C21H19ClN2O4 | 398.843 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62669-70-9
Molecular Formula C21H19ClN2O4
Molecular Weight (g/mol) 398.843
MDL Number MFCD00012664
InChI Key SEXIURXGNASJRB-UHFFFAOYSA-N
PubChem CID 16211830
IUPAC Name methyl 2-(3-amino-6-iminoxanthen-9-yl)benzoate;hydrate;hydrochloride
SMILES COC(=O)C1=CC=CC=C1C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N.O.Cl

Specifications

Assay Percent Range 99+%
Merck Index 15, 8306
Infrared Spectrum Authentic
Formula Weight 380.82
Grade Extra Pure
Packaging Glass bottle
UV_Vis 99.0% min.
Color Brown to Brown/Red or Green
Quantity 100 mg
Physical Form Powder
Chemical Name or Material Rhodamine 123, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Rhodamine 123
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.