missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 6G

Product Code. 15484719
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484719 100 g 100g
15474719 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484719 Supplier Thermo Scientific Chemicals Supplier No. J62315.22
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for fluorescent labelling | CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 989-38-8
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.017
MDL Number MFCD00012665
InChI Key VYXSBFYARXAAKO-UHFFFAOYSA-N
Synonym Basic Red 1; C.I. 45160
PubChem CID 51358423
IUPAC Name ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
SMILES [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]

Specifications

Beilstein 3900071
Color Brown to Red
Solubility Information Soluble in water.
Quantity 100 g
Physical Form Powder
Formula Weight 479.02
Chemical Name or Material Rhodamine 6G

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.