missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Rhodamine 6G

Product Code. p-200104503
Encompass_Preferred
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 16482419

Brand: MedChemExpress HYD0309

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rhodamine 6G is a rhodamine analog useful in Pgp efflux assays. It can be used in characterizing the kinetics of MRP1- mediated efflux, and as a laser dye and potential mitochondrial probe.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 1 g
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (52.19 mM; Need ultrasonic) ∣ H2O : 10 mg/mL (20.88 mM; ultrasonic and warming and heat to 60°C)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 479.01 g/mol
CAS 989-38-8
Synonym Basic Red 1
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.