missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 6G, 99%, pure

Product Code. 10147990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10147990 5 g Glass bottle 5g
10045820 25 g Glass bottle 25g
10587362 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147990 Supplier Thermo Scientific Chemicals Supplier No. 419010050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 989-38-8
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.017
MDL Number MFCD00012665
InChI Key VYXSBFYARXAAKO-UHFFFAOYSA-N
Synonym Basic Red 1, C.I. 45160
PubChem CID 51358423
IUPAC Name ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
SMILES [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]

Specifications

Melting Point 290.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
UV_Vis 98.5% min.
Beilstein 18, V,12, 283
Color Brown to Brown/Red
Solubility Information Solubility in water: soluble.
Quantity 5 g
Physical Form Fine Crystalline Powder
Formula Weight 479.01
Percent Purity 99%
Grade Pure
Chemical Name or Material Rhodamine 6G, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Rhodamine 6G
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.