missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine 6G, TRC

Product Code. 30419381
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419381 1 g
30416121 5 g
30415142 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419381 Supplier Toronto Research Chemicals Supplier No. TRCR3185951G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 989-38-8
Molecular Formula C28 H30 N2 O3 . Cl H
Molecular Weight (g/mol) 479.01
Synonym Benzoic acid, o-[6-(ethylamino)-3-(ethylimino)-2,7-dimethyl-3H-xanthen-9-yl]-, ethyl ester, monohydrochloride (8CI), Rhodamine 6GCP (6CI), Xanthylium, 9-[2-(ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethyl-, chloride (9CI), 6-Carboxyrhodamine, Aizen Rhodamine 6GCP, Basic Red 1, Basonyl Red 482, Basonyl Red 483, C.I. 45160, C.I. Basic Red 1, Calcozine Red 6G, Calcozine Rhodamine 6GX, Eljon Pink Toner, Exciton 590, Fanal Pink B, Fanal Pink GFK, Fanal Red 25532, Flexo Red 482, Heliostable Brilliant Pink B extra, Mitsui Rhodamine 6GCP, NSC 36345, Nyco Liquid Red GF, R 634, R 6G, Rh 6G, Rhodamin 6G, Rhodamine 4GD, Rhodamine 4GH, Rhodamine 5GDN, Rhodamine 5GDN Extra, Rhodamine 5GL, Rhodamine 6G, Rhodamine 6G Extra, Rhodamine 6G Extra Base, Rhodamine 6G chloride, Rhodamine 6GB, Rhodamine 6GBN, Rhodamine 6GD, Rhodamine 6GDN, Rhodamine 6GDN Extra, Rhodamine 6GEx ethyl ester, Rhodamine 6GH, Rhodamine 6GO, Rhodamine 6GX, Rhodamine 6JH, Rhodamine 6JH-SA, Rhodamine 6JH-SA Extra 1150, Rhodamine 6Zh-DN, Rhodamine F 5G, Rhodamine F 5GL, Rhodamine GDN, Rhodamine GDN Extra, Rhodamine Y 20-7425, Rhodamine Zh, Silosuper Pink B, Vali Fast Red 1308
IUPAC Name ethyl 2-[(6E)-3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydrochloride
SMILES Cl.CCNc1cc2OC3=C\C(=N/CC)\C(=CC3=C(c4ccccc4C(=O)OCC)c2cc1C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H/b30-24+;
Formula Weight 478.2023
Percent Purity >95%
Chemical Name or Material Basic Red 1
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.