missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine B, TRC

Product Code. 30420913
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420913 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420913 Supplier Toronto Research Chemicals Supplier No. TRCR318605250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rhodamine B chloride
CAS 81-88-9
Molecular Formula C28 H31 N2 O3 . Cl
Quantity 250 g
Synonym Rhodamine B, Xanthylium','9-(2-carboxyphenyl)-3,6-bis(diethylamino)-','chloride (1:1), Ammonium','[9-(o-carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene]diethyl-','chloride (8CI), Xanthylium','9-(2-carboxyphenyl)-3,6-bis(diethylamino)-','chloride (9CI), [9-(o-Carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene]diethylammonium chloride (6CI,7CI), 11411 Red, ADC Rhodamine B, Aizen Rhodamine B, Aizen Rhodamine BH, Aizen Rhodamine BHC, Akiriku Rhodamine B, Amezin Rhodamine B Liquid, Basazol Red 71L, Basazol Red 71P, Basic Rose Extract, Basic Rose Red, Basic Violet 10, Basonyl Red 540, Basonyl Red 545, Basonyl Red 545FL, Brilliant Pink B, C.I. 45170, C.I. Basic Violet 10, C.I. Food Red 15, Calcozine Red BX, Calcozine Rhodamine BXP, Cerise Toner X 1127, Chromatint Rhodamine B, D and C Red No. 19, D&C Red 19, D&C Red No. 19, Diabasic Rhodamine B, Edicol Supra Rose B, Edicol Supra Rose BS, Eriosin Rhodamine B, FD And C Red No. 19, Flexo Red 540, GRM 980, Hexacol Rhodamine B Extra, Ikada Rhodamine B, Japan Red 213, Japan Red No. 213, LC 6100, Mitsui Rhodamine BX, OP 312, RHB, Red No. 213, Rheonine B, Rhodamine 610 chloride, Rhodamine B, Rhodamine B 500, Rhodamine B 500 hydrochloride, Rhodamine B Extra, Rhodamine B Extra M 310, Rhodamine B Extra S, Rhodamine B-EX, Rhodamine BA, Rhodamine BA Export, Rhodamine BDC, Rhodamine BN, Rhodamine BS, Rhodamine BX, Rhodamine BXL, Rhodamine BXP, Rhodamine FB, Rhodamine Lake Red B, Rhodamine O, Rhodamine S, Rhodamine S (Russian), Rhodamine','tetraethyl-, Sicilian Cerise Toner A 7127, Sumiplast Pink B, Symulex Magenta F, Symulex Rhodamine B Toner F, Takaoka Rhodamine B, Tetraethylrhodamine
InChI Formula InChI=1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H
SMILES [Cl-].CCN(CC)c1ccc2C(=C3C=CC(=[N+](CC)CC)C=C3Oc2c1)c4ccccc4C(=O)O
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
Molecular Weight (g/mol) 479.01
Formula Weight 478.2023
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.