missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodamine B, TRC
Specifications
Specifications
| Chemical Name or Material | Rhodamine B chloride |
| CAS | 81-88-9 |
| Molecular Formula | C28 H31 N2 O3 . Cl |
| Quantity | 500 g |
| Synonym | Rhodamine B, Xanthylium','9-(2-carboxyphenyl)-3,6-bis(diethylamino)-','chloride (1:1), Ammonium','[9-(o-carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene]diethyl-','chloride (8CI), Xanthylium','9-(2-carboxyphenyl)-3,6-bis(diethylamino)-','chloride (9CI), [9-(o-Carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene]diethylammonium chloride (6CI,7CI), 11411 Red, ADC Rhodamine B, Aizen Rhodamine B, Aizen Rhodamine BH, Aizen Rhodamine BHC, Akiriku Rhodamine B, Amezin Rhodamine B Liquid, Basazol Red 71L, Basazol Red 71P, Basic Rose Extract, Basic Rose Red, Basic Violet 10, Basonyl Red 540, Basonyl Red 545, Basonyl Red 545FL, Brilliant Pink B, C.I. 45170, C.I. Basic Violet 10, C.I. Food Red 15, Calcozine Red BX, Calcozine Rhodamine BXP, Cerise Toner X 1127, Chromatint Rhodamine B, D and C Red No. 19, D&C Red 19, D&C Red No. 19, Diabasic Rhodamine B, Edicol Supra Rose B, Edicol Supra Rose BS, Eriosin Rhodamine B, FD And C Red No. 19, Flexo Red 540, GRM 980, Hexacol Rhodamine B Extra, Ikada Rhodamine B, Japan Red 213, Japan Red No. 213, LC 6100, Mitsui Rhodamine BX, OP 312, RHB, Red No. 213, Rheonine B, Rhodamine 610 chloride, Rhodamine B, Rhodamine B 500, Rhodamine B 500 hydrochloride, Rhodamine B Extra, Rhodamine B Extra M 310, Rhodamine B Extra S, Rhodamine B-EX, Rhodamine BA, Rhodamine BA Export, Rhodamine BDC, Rhodamine BN, Rhodamine BS, Rhodamine BX, Rhodamine BXL, Rhodamine BXP, Rhodamine FB, Rhodamine Lake Red B, Rhodamine O, Rhodamine S, Rhodamine S (Russian), Rhodamine','tetraethyl-, Sicilian Cerise Toner A 7127, Sumiplast Pink B, Symulex Magenta F, Symulex Rhodamine B Toner F, Takaoka Rhodamine B, Tetraethylrhodamine |
| InChI Formula | InChI=1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H |
| SMILES | [Cl-].CCN(CC)c1ccc2C(=C3C=CC(=[N+](CC)CC)C=C3Oc2c1)c4ccccc4C(=O)O |
| IUPAC Name | [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride |
| Molecular Weight (g/mol) | 479.01 |
| Formula Weight | 478.2023 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Vous avez repéré une opportunité d'amélioration ?