missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine WT, 20% solution in water

Product Code. 12193761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12193761 1 L 1 L
12103771 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12193761 Supplier Thermo Scientific Chemicals Supplier No. 446970010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37299-86-8 | C29H29ClN2Na2O5 | 566.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rhodamine WT
CAS 37299-86-8, 7732-18-5
Packaging Glass bottle
Specific Gravity 1.16
Molecular Formula C29H29ClN2Na2O5
Density 1.1600g/mL
Solubility Information Solubility in water: miscible.
Quantity 1 L
InChI Key NRZDMKVYRRMFRR-UHFFFAOYSA-L
SMILES CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)C(=O)[O-])C(=O)[O-].[Na+].[Na+].[Cl-]
IUPAC Name disodium;4-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzene-1,3-dicarboxylate;chloride
Molecular Weight (g/mol) 566.99
PubChem CID 37718
Formula Weight 566.99
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.