missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) trifluoroacetate dimer

Product Code. 10771894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771894 1 g Glass bottle 1g
10568833 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10771894 Supplier Thermo Scientific Acros Supplier No. 389700010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31126-95-1 | C8F12O8Rh2 | 657.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31126-95-1
Molecular Formula C8F12O8Rh2
Molecular Weight (g/mol) 657.88
MDL Number MFCD00209611
InChI Key SZQVMUPTZFMHQT-UHFFFAOYSA-N
Synonym rhodium ii trifluoroacetate dimer, trifluoroacetic acid rhodium ii salt dimer, dirhodium ii tetra trifluoroacetate, rhodium ii trifluoroacetate dimer, ?rh2 cf3coo 4, bis 2,2,2-trifluoroacetyl oxy rhodio 2,2,2-trifluoroacetate
PubChem CID 10985187
IUPAC Name rhodium;2,2,2-trifluoroacetic acid
SMILES C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O.[Rh].[Rh]

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 657.88
Chemical Name or Material Rhodium(II) trifluoroacetate dimer
compliance-icons
Product Identifier
  • Rhodium(II) trifluoroacetate dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.