missing translation for 'onlineSavingsMsg'
Learn More

Rhodizonic Acid Sodium Salt, Pure, Fisher Chemical

Product Code. 10132350 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132350 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10132350 Supplier Fisher Chemical Supplier No. R/0300/44
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 523-21-7
Color Green
Physical Form Powder/Solid
Quantity 5 g
Molecular Formula C6Na2O6
MDL Number MFCD00001650
Synonym sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
InChI Key SXWPCWBFJXDMAI-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O
IUPAC Name disodium tetraoxocyclohex-1-ene-1,2-bis(olate)
Molecular Weight (g/mol) 214.04
PubChem CID 68225
Formula Weight 214.04g/mol
Packaging Glass Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.