missing translation for 'onlineSavingsMsg'
Learn More

Rhynchophylline, MedChemExpress

Product Code. 16428968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428968 10 mg 10mg
16411398 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428968 Supplier Medchem Express Supplier No. HYN0387

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rhyncholphylline, an alkaloid isolated from Uncaria, shows potent inhibition of lipopolysaccharide (LPS)-induced NO production in rat primary microglial cells.

Rhyncholphylline, an alkaloid isolated from Uncaria, shows potent inhibition of lipopolysaccharide (LPS)-induced NO production in rat primary microglial cells.

Specifications

Chemical Name or Material Rhynchophylline
CAS 76-66-4
Color White
Purity Grade Notes Research
Molecular Formula C22H28N2O4
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (65.02 mM; Need ultrasonic)
SMILES O=C(NC1=C2C=CC=C1)[C@]32[C@@](C[C@H](/C(C(OC)=O)=C\OC)[C@@H](CC)C4)([H])N4CC3
Molecular Weight (g/mol) 384.47
Formula Weight 384.47
Percent Purity 98.51%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.