missing translation for 'onlineSavingsMsg'
Learn More

Ribavirin, MP Biomedicals™

Product Code. 11462701 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462701 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11462701 Supplier MP Biomedicals Supplier No. 0219606683
  • ONLINE EXCLUSIVE PRICE
    1115.00 €
    1056.30 € / 250mg
    Save 58.70 €
    5% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Active against many DNA and RNA viruses. Ribavirin, MP Biomedicals is an antiviral compound that is active against RSV, HIV, HCV and other infectious agents. It is a nucleoside antimetabolite drug that interferes with duplication of viral genetic material

Ribavirin is used as an antiviral agent against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. It inhibits purine (GTP pools) biosynthesis and is a precursor to the di- and tri-phosphates that inhibit viral RNA-dependent RNA polymerases. Its metabolite, ribavirin 5'-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase, but many other mechanisms of action are also supported with experimental evidence. It is also an analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.207
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
Synonym ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
PubChem CID 37542
ChEBI CHEBI:63580
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N

Specifications

Melting Point 173°C (literature)
Loss on Drying 0.04% (Lit.)
Quantity 250 mg
Solubility Information Soluble in water (20mg/mL, clear, colorless solution).
Formula Weight 244.2g/mol
Percent Purity >99%
Physical Form Powder
compliance-icons
Product Identifier
  • Ribavirin
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ribavirin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.