missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin 3',5'-Diphosphate Ammonium Salt (>90%), TRC

Product Code. 30424269
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424269 5 mg 5mg
30411370 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424269 Supplier Toronto Research Chemicals Supplier No. TRCR4150305MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Riboflavin 3',5'-Diphosphate Ammonium Salt
Molecular Formula C17H22N4O12P2 • x(NH3)
Quantity 5 mg
Synonym Benzo[g]pteridine Riboflavin 3'-(Dihydrogen Phosphate) Derivative Ammonium Salt
InChI Formula InChI=1S/C17H22N4O12P2.H3N/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(24)20-17(25)19-15)5-11(22)14(33-35(29,30)31)12(23)6-32-34(26,27)28;/h3-4,11-12,14,22-23H,5-6H2,1-2H3,(H,20,24,25)(H2,26,27,28)(H2,29,30,31);1H3/t11-,12+,14-;/m0./s1
SMILES O=C(C1=NC2=CC(C)=C(C)C=C2N(C[C@@H](O)[C@@H](OP(O)(O)=O)[C@H](COP(O)(O)=O)O)C1=N3)NC3=O
IUPAC Name (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,4-dihydroxypentane-1,3-diyl bis(dihydrogen phosphate), ammonia salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.