missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin 5'-(Dihydrogen Phosphate) Monosodium Salt Dihydrate (~70%), TRC

Product Code. 30412021
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412021 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412021 Supplier Toronto Research Chemicals Supplier No. TRCR414990100MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Riboflavin-5'-phosphate
CAS 6184-17-4
Molecular Formula C17 H20 N4 O9 P . Na . 2 H2 O
Quantity 100 mg
Synonym Riboflavin sodium phosphate dihydrate, Riboflavin 5'-(dihydrogen phosphate)','sodium salt','hydrate (1:1:2), Riboflavin 5'-(dihydrogen phosphate)','monosodium salt','dihydrate (9CI), Riboflavine 5'-(dihydrogen phosphate)','monosodium salt','dihydrate (8CI), Benzo[g]pteridine','riboflavin 5'-(dihydrogen phosphate) deriv., Alloxazine mononucleotide sodium salt dihydrate, Coflavinase dihydrate, Cytoflav dihydrate, Flavin mononucleotide sodium salt dihydrate, Riboflavin 5'-phosphate monosodium salt dihydrate, Riboflavin 5'-phosphate sodium dihydrate, Vitamin B2 phosphate (sodium salt) dihydrate
InChI Formula InChI=1S/C17H21N4O9P.Na.2H2O/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;;;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);;2*1H2/q;+1;;/p-1/t11-,12+,14-;;;/m0.../s1
SMILES O.O.[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(=O)(O)[O-])c2cc1C
IUPAC Name sodium;[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] hydrogen phosphate;dihydrate
Molecular Weight (g/mol) 514.36
Formula Weight 514.1077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.