missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin, 97%, Electrophoresis Grade

Product Code. 15448069
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448069 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15448069 Supplier Thermo Scientific Alfa Aesar Supplier No. J66632.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83-88-5
Melting Point 290°C
Color Orange-Yellow
Odor Odorless
Molecular Formula C17H20N4O6
MDL Number MFCD00005022
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,8200
Synonym riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
IUPAC Name 7,8-dimethyl-10-(2,3,4,5-tetrahydroxypentyl)-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
Molecular Weight (g/mol) 376.37
PubChem CID 71310809
Formula Weight 376.37
Percent Purity 98%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material Riboflavin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.