missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin USP Grade, MP Biomedicals™

Product Code. 11496192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496192 1 kg 1kg
11475985 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496192 Supplier MP Biomedicals Supplier No. 0210281391
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

USP Grade Orange Powder

  • Vitamin/enzyme cofactor found in milk, eggs, malted barley, liver, kidney, heart, leafy vegetables
  • Used as a photoinitiator for polymerization of polyacrylamide gels

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
Synonym Vitamin B2, Lactoflavin, Vitaming, 7,8-Dimethyl-10- (D-ribo-2,3,4,5- tetrahydroxypentyl) isoalloxazine
PubChem CID 71310809
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point 290°C (decomposition)(literature)
Color Orange
Loss on Drying ≤1.5%
Quantity 1 kg
Beilstein 97825
Solubility Information Soluble in 0.1 M Sodium Hydroxide (50mg/mL - clear, yellowish-orange solution); only slightly soluble in water.
Optical Rotation (c = 0.5, 0.05 M NaOH) -115° to -135°
Physical Form Powder
Assay >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.