missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RiboMinus™ Bacteria 2.0 Transcriptome Isolation Kit

Product Code. 17173239
Change view
Click to view available options
Quantity:
12 reactions
Unit Size:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17173239 12 reactions 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17173239 Supplier Invitrogen™ Supplier No. A47335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The RiboMinus Bacteria 2.0 Transcriptome Isolation Kit is a complete and automatable kit that enables efficient and selective removal of bacterial and archaebacterial ribosomal RNA (rRNA) from intact total RNA (100 ng to 5 μg) in less than one hour.

Highly efficient depletion of 16S, 23S, and 5S rRNA from a variety of species

The RiboMinus Bacteria 2.0 Transcriptome Isolation Kit is a complete and automatable kit that enables efficient and selective removal of bacterial and archaebacterial ribosomal RNA (rRNA) from intact total RNA (100 ng to 5 μg) in less than one hour.

Key features of the RiboMinus Bacteria 2.0 kit:

  • Efficient—the included RiboMinus Pan-Prokaryote Probe Mix has consistently shown less than 10% of reads mapping to 16S and 23S rRNA reads
  • Wide range of targets—designed with >150 probe designs targeting 16S, 23S, and 5S rRNA targets from >76 representative species of bacteria and archaebacteria from common phyla and sub-phyla
  • Convenient—Dynabeads MyOne Streptavidin C1 magnetic beads and bead cleanup module and protocols included for Kingfisher instruments
  • Flexible— accommodates mass inputs from 100 ng to 5 μg of total RNA; works with wide range of total RNA samples with RIN values from 5.0–10.0
  • High detection sensitivity—increases the number of sequencing reads mapping to protein-coding transcripts compared to previous version
  • Suitable for metatranscriptomics—deplete rRNA of many microbial species in metagenomic samples

Target rRNA from panorama of bacteria and archaebacteria

The RiboMinus Bacteria 2.0 kit is based on hybridization-capture technology and includes a new RiboMinus Pan-Prokaryote Probe Mix, which results in a 4-fold decrease of ribosomal reads remaining and shows significant improvement over the previous bacterial probe mix. The new probe mix consists of over 150 3’-biotin-labeled ssDNA oligonucleotiode designs. These designs were created using highly conserved regions of rRNA across 76 pan-prokaryote species (see table below) and tested with several species (see compatibility list below).

Dynabeads MyOne Streptavidin C1 cleanup kit included

Once the probe mix has selectively hybridized to the target rRNA transcripts, removal of the probe-rRNA complex is completed by capture on Dynabeads MyOne Streptavidin C1 magnetic beads, leaving behind rRNA-depleted RNA. The RiboMinus Bacteria 2.0 kit includes a magnetic bead-based cleanup module for concentration of the rRNA-depleted RNA, enabling easier handling, scalability, and automation on KingFisher instruments. The resulting rRNA-depleted RNA (RiboMinus RNA) is suitable for whole-transcriptome analysis by RNA-Seq or other downstream applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage RiboMinus Isolation Kits include: RiboMinus Magnetic Beads, RiboMinus human/mouse, bacteria, or yeast probe, and Hybridization Buffer

The optional RiboMinus Concentration Module contains spin cartridges, wash tubes, recovery tubes, Binding Buffer, Wash Buffer, and RNase-free (DEPC-treated) water.

Store the isolation modules at 4°C and the concentration module at room temperature. All reagents are guaranteed stable for 6 months when properly stored.
Isolation Technology Magnetic Bead
Sample Type Total RNA (Bacterial)
Final Product Type Transcriptome RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Next Generation Sequencing, Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction
No. of Reactions 12 reactions
Product Line RiboMinus
Quantity 12 reactions
Starting Material Amount 100 ng to 5 ug of total RNA
Target Transcriptome RNA
System Type RiboMinus
Form Magnetic Bead
Show More Show Less

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.