missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RiboMinus™ Pan-Prokaryote Probe Mix

Product Code. 17125792
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17125792 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17125792 Supplier Invitrogen™ Supplier No. A46920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The RiboMinus™ Pan-Prokaryote Probe Mix is a pool of more than 150 3'-biotin labeled ssDNA oligonucleotides that are used to capture and eliminate bacterial and archaebacterial ribosomal RNA from RNA samples in preparation for NGS transcriptome studies.

RiboMinus™ Pan-Prokaryote Probe Mix is a component of the RiboMinus Bacteria 2.0 Transcriptome Isolation Kit and is available as a standalone product for use in conjunction with other RiboMinus products targeting other species: Plant (Cat. No. A1083808) and Eukaryote v2 (Cat. No. A15026). Combining RiboMinus Pan-Prokaryote Probe Mix with Plant or Eukaryote probe mixes is useful for metatranscriptomic studies. This probe mix consists of over 150 3’-biotin-labeled ssDNA oligonucleotide designs. These designs were created using highly conserved regions of rRNA across 76 pan-prokaryote species (see table below) and tested with several species (see compatibility list below).
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage RiboMinus Isolation kits include RiboMinus Magnetic Beads, RiboMinus human/mouse, bacteria, or yeast probe, and Hybridization Buffer. The optional RiboMinus Concentration Module contains spin cartridges, wash tubes, recovery tubes, Binding Buffer, Wash Buffer, and RNase-free (DEPC-treated) water. Store the isolation modules at +4°C and the concentration module at room temperature. All reagents are guaranteed stable for 6 months when properly stored.
Isolation Technology Magnetic Bead
Purification Time 1.25 hr.
Sample Type Total RNA (Bacterial)
Final Product Type Transcriptome RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Next Generation Sequencing, Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction
No. of Reactions 12
Product Line RiboMinus
Quantity 1
Starting Material Amount 100 ng to 5 ug of total RNA
Target Transcriptome RNA
System Type RiboMinus
Form Magnetic Bead
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.