missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Richard-Allan Scientific™ Iron Stain, Nuclear Red Thiazin stain solution

Product Code. 10117998 Shop All Epredia Products
Click to view available options
Type:
Hydrochloric Acid Solution (20%)
Iron Stain Kit
Nuclear Fast Red
Potassium Ferrocyanide
Unit Size:
125mL
250mL
Each
This item is not returnable. View return policy

Product Code. 10117998

Brand: Epredia™ 87006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Richard-Allan Scientific™ Iron Stain.

  • Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
  • Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink
TRUSTED_SUSTAINABILITY

Specifications

Type Iron Stain Kit
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • H371-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P313-Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P234-Keep only in original container.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P363-Wash contaminated clothing before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/polypropylene container with a resistant inner liner.
  • P402-Store in a dry place.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, potassium ferrocyanide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.