Learn More
Epredia™ Modified Mayers Hematoxylin
Produce crisp nuclear staining with Epredia™ Richard-Allan Scientific™ Modified Mayer's Hematoxylin, a progressive nuclear stain.
Brand: Epredia™ 72804
130.15 EUR valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Code : A0
Additional Details : Weight : 1.88000kg
Description
- Difficult to overstain with product
- Requires no acid rinse and does not stain mucin
- Recommended for use when less intense results are desired
- An effective counterstain in immunoperoxidase and special stain procedures
Specifications
Hematoxylin | |
1 pt. |