Learn More
Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y 7111
Create an ideal visual contrast to Hematoxylin with the fast-acting Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y counterstain.
Brand: Epredia™ 7111
Code : NEW
Additional Details : CAS Number : 64-17-5 Weight : 2.26800kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- Bright, vivid cytoplasmic staining
- Creates three distinct shades of pink
- Unmatched color contrast between the nuclei and cytoplasm
- Various shades of red and pink differentiate muscle, red blood cells and connective tissue
- Strict quality assurance guarantees product consistency and longevity
- Quantitative testing eliminates need for adjustments in dilution or time
Available in 5 liter packaging
- Reduces plastic by 90%
- Reduces stain spills
- Increases stain stability
- Saves 35% storage space versus traditional packaging
Specifications
Eosin Stain | |
1 pt. |