missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Pen-Fix™

Product Code. 15337195 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 5 L
4 x 1 gal.
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15337195 4 x 5 L Each
10148438 4 x 1 gal. Pack of 4
13338656 5 gal. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15337195 Supplier Epredia™ Supplier No. 6115
  • ONLINE EXCLUSIVE PRICE
    824.88 €
    653.63 € / Pack of 1
    Save 171.25 €
    20% Off
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.

  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 x 5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.